Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell RSC PureFood GMO and Authentication Kit, 48 preps

Catalog No. PRAS1600 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Maxwell RSC PureFood GMO and Authentication Kit, 48 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRAS1600 Maxwell RSC PureFood GMO and Authentication Kit, 48 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRAS1600 Supplier Promega Supplier No. AS1600
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Maxwell RSC PureFood GMO and Authentication Kit used with the Maxwell RSC Instrument provides an easy, automated method for efficient purification of DNA used in PCR-based testing for GMO DNA sequences and PCR-based food and ingredient authentication.

  • Safe, Automated DNA Purification from Food and Feed Samples
  • Purifies high−quality DNA from a range of food and feed samples
  • Results in highly concentrated DNA that is ready to use in downstream assays
  • Simple, five−step protocol saves time and eliminates organic extraction steps

Specifications

Content And Storage 15°C to 30°C
Format Kit
Isolation Technology Magnetic Bead
No. of Reactions 48 Preps
Product Type DNA Purification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.