Learn More
Promega Maxwell™ RSC simplyRNA Blood Kit
Catalog No. PRAS1380 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
48 Preps
Catalog No. Quantity
PRAS1380 48 Preps
1 options
Catalog No. PRAS1380 Supplier Promega Supplier No. AS1380
The Maxwell RSC simplyRNA Blood Kit, used with the Maxwell RSC Instrument, provides a simple method for automated RNA purification from fresh whole blood. The Maxwell RSC Instrument can process from 1 to 16 samples in under an hour.
  • Automated Purification of RNA from Whole Blood using a Low Elution Volume
  • Purifies total RNA from 2.5mL of fresh whole blood
  • Reduces centrifugation steps
  • Yields highly concentrated RNA
Specifications
Content And Storage 15°C to 30°C
Quantity 48 Preps