Learn More
Promega Maxwell™ RSC simplyRNA Tissue Kit
Catalog No. PRAS1340 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
48 Preps
Catalog No. Quantity
PRAS1340 48 Preps
1 options
Catalog No. PRAS1340 Supplier Promega Supplier No. AS1340
The Maxwell RSC simplyRNA Tissue Kit, used with the Maxwell RSC Instrument, provides a simple method for automated RNA purification from up to 20mg tissue samples. The Maxwell RSC Instrument can process from 1 to 16 samples in under an hour.
  • Automated RNA Purification from Tissues using a Low Elution Volume
  • Purifies total RNA from up to 20mg of tissue
  • Reduces pre-extraction sample handling to 4 steps
  • No beta-mercaptoethanol needed
Specifications
Content And Storage 15°C to 30°C
Quantity 48 Preps