Learn More
Description
- Automated DNA Purification from Stabilized Saliva Samples
- Simple protocol with optimized reagents
- Consistent DNA yields
- DNA ready to use in downstream assays such as HLA typing
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| No. of Reactions | 48 Preps |
| Product Type | DNA Purification Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.