Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell RSC Tissue DNA Kit, 48 preps

Catalog No. PRAS1610 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Maxwell RSC Tissue DNA Kit, 48 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRAS1610 Maxwell RSC Tissue DNA Kit, 48 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRAS1610 Supplier Promega Supplier No. AS1610
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Maxwell RSC Tissue DNA Kit is a simple, automated method for extraction of DNA from up to 50mg of mammalian tissue using the Maxwell RSC Instrument.

  • Automated DNA Extraction from Mammalian Tissue
  • Extracts DNA from up to 50mg of mammalian tissue
  • Purifies high yields of amplifiable DNA
  • Automated protocol improves efficiency

Specifications

Content And Storage 15°C to 30°C
Format Kit
No. of Reactions 48 Preps
Product Type DNA Purification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.