Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell RSC Whole Blood DNA Kit, 48 preps

Catalog No. PRAS1520 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Maxwell RSC Whole Blood DNA Kit, 48 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRAS1520 Maxwell RSC Whole Blood DNA Kit, 48 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRAS1520 Supplier Promega Supplier No. AS1520
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Maxwell RSC Whole Blood DNA Kit is a simple, automated method for extraction of DNA from 50-500ul of whole blood samples using the Maxwell RSC Instrument. No preprocessing is necessary; purified DNA is ready for analysis in less than 40 minutes.

  • Automated DNA Purification from Whole Blood
  • Purify DNA from 50−500μl of whole blood in less than 40 minutes with no preprocessing
  • Yields highly pure DNA without PCR inhibitors to ensure success in sensitive downstream applications
  • Flexible elution volume lets you maximize concentration or yield

Specifications

Content And Storage 15°C to 30°C
Format Kit
Isolation Technology Magnetic Bead
No. of Reactions 48 Preps
Product Type DNA Purification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.