Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ MAXX Pack Absorbent Pack

Catalog No. 19103118 Shop All Brady Corporation Products
Change view
Click to view available options
Type:
Spill Kit, Oil-only
Universal
Color:
Gray
White
For Use With (Application):
Oil-only spills
Spills
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Color For Use With (Application)
19-103-118 Spill Kit, Oil-only Gray Oil-only spills
19-103-119 Universal White Spills
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-103-118 Supplier Brady™ Supplier No. MXPO20

Avoid slips, trips, and falls with this great addition to your absorbent program. The Brady™ MAXX Pack Absorbent pack absorbs most liquids.

  •  Economic solution for oil spill containment
  • Pack contains 20 full-size, single-ply absorbent pads in a re-sealable zipper bag
  • Great for storing around facilities for immediate spill response

Specifications

Absorbancy 18 gl/gs
Color Gray
Capacity (English) 35 gal.
Material Polypropylene
Type Spill Kit, Oil-only
For Use With (Application) Oil-only spills
Packaging Quantity 5/Pk., 4 Pk./Cs.
Dimensions (L x W) 15 x 19 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.