Learn More
Description
- Economic solution for spill containment
- Pack contains 20 full-size, single-ply absorbent pads in a re-sealable zipper bag
- Great for storing around facilities for immediate spill response
Specifications
Specifications
| Absorbancy | 18 gl/gs |
| Color | White |
| Capacity (English) | 35 gal. |
| Material | Polypropylene |
| Type | Universal |
| For Use With (Application) | Spills |
| Packaging Quantity | 5/Pk., 4 Pk./Cs. |
| Dimensions (L x W) | 15 x 19 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.