Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Maxymum Recovery™ 200 μL Universal Pipetter Tips: 200 μL, Beveled

Catalog No. 14222714 Shop All Axygen Products
Change view
Click to view available options
Format:
Bulk
Hinged Rack
Stacked Rack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
14-222-714 Hinged Rack
14-222-713 Bulk
14-222-716 Stacked Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-222-714 Supplier Axygen™ Supplier No. T200CLR
Add to Cart
Add to Cart

Designed to fit a wide variety of single and multi-channel pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model T-200-C-L-R
Certifications/Compliance DNA free, RNase free, Endotoxin free
Clean Claims DNase, RNase, DNA, and PCR inhibitor Free
Format Hinged Rack
Tip Style Beveled, Low Retention, Universal
Packaging Hinged rack; 960/Pk, 4800/Cs
Quantity 96 Tips/Rack, 960/Pk., 4800/Cs.
Product Type Universal Fit Pipette Tips
Includes Tips, Rack, Pack, Case
For Use With (Equipment) Universal Fit
Product Line Maxymum Recovery
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.