Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

May-Grünwald's Eosine-Methylene Blue Solution Modified, For Microscopy, MilliporeSigma™

Catalog No. M1014240100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
25 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1014240100 100 mL
M1014240500 500 mL
M1014241000 1 L
M1014242500 2.5 L
M1014249025 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. M1014240100 Supplier MilliporeSigma Supplier No. 1014240100
Add to Cart
Add to Cart
For microscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material May-Grünwald's Eosine-Methylene Blue Solution Modified
UN Number UN 1230
Odor Smells like methanol
Packaging Glass bottle
Vapor Pressure 125 hPa at 20°C
Color Blue
Quantity 100 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.