Learn More
May-Grünwald's Eosine-Methylene Blue Solution Modified, For Microscopy, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Chemical Name or Material | May-Grünwald's Eosine-Methylene Blue Solution Modified |
| UN Number | UN 1230 |
| Odor | Smells like methanol |
| Packaging | Plastic bottle |
| Vapor Pressure | 125 hPa at 20°C |
| Color | Blue |
| Quantity | 2.5 L |
| Physical Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.