Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals May-Gruenwald Stain, pure

Catalog No. AC194830250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC194830250 25 g
AC194831000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC194830250 Supplier Thermo Scientific Chemicals Supplier No. 194830250
Add to Cart

CAS: 524698-43-9

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material May-Gruenwald Stain
CAS 524698-43-9
MDL Number MFCD00131580
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Lambda Max in 66:33 IMS:water, 655 to 670nm (blue), 515 to 530nm (red)
Grade Pure
Packaging Glass bottle
Color Brown or Green
Quantity 25 g
Physical Form Fine Crystalline Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.