Learn More
Stoelting™ Mayo-Noble Scissors
SDP
Catalog No. 10001718 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved
Straight
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-718 Straight
10-001-719 Curved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-001-718 Supplier Stoelting™ Supplier No. 5213668
Only null left
Add to Cart
Add to Cart
Curved or straight scissors
  • Straight-bladed style is designed for cutting body tissues near the surface of a wound.
  • Curved-bladed styles allow deeper penetration into the wound and are also used to cut thick tissues
Specifications
For Use With (Application) Neuroscience
Length (English) 6.5 in.
Model Straight