Learn More
Stoelting™ Mayo Scissors
SDP
Catalog No. 10001712 Shop All Stoelting Co Products
Change view
Click to view available options
Length (English):
5.5 in.
5.75 in.
6.75 in.
9 in.
Model:
Curved
Curved Carbide
Curved, Surecut
Curved, Tungsten carbide
Straight
Straight Carbide
Straight, Left-handed
Straight, Surecut
Straight, Tungsten carbide
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Model
10-001-712 6.75 in. Straight Carbide
10-001-703 5.5 in. Curved, Surecut
10-001-701 5.5 in. Straight, Surecut
10-000-544 5.5 in. Curved, Tungsten carbide
10-000-543 5.5 in. Straight, Tungsten carbide
10-000-541 5.75 in. Curved
10-000-540 5.75 in. Straight
10-001-707 6.75 in. Curved
10-001-713 6.75 in. Curved Carbide
10-001-720 6.75 in. Curved, Surecut
10-001-704 6.75 in. Straight
10-001-706 6.75 in. Straight, Surecut
10-000-542 6.75 in. Straight, Left-handed
10-001-709 9 in. Curved, Surecut
10-001-708 9 in. Straight, Surecut
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 10-001-712 Supplier Stoelting™ Supplier No. 5213662
Add to Cart
Add to Cart
  • Straight-bladed style is designed for cutting body tissues near the surface of a wound.
  • Curved-bladed styles allow deeper penetration into the wound and are also used to cut thick tissues
Specifications
For Use With (Application) Neuroscience
Length (English) 6.75 in.
Model Straight Carbide