Learn More
Medchemexpress LLC MAYTANSINE 5MG
SDP
Supplier:  Medchemexpress LLC HY136745MG
Encompass_Preferred
Maytansine is a highly potent microtubule-targeted compound that induces mitotic arrest and kills tumor cells at subnanomolar concentrations.cas no: 35846-53-8
Catalog No. NC2203328
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.