Learn More
Invitrogen™ MAZ Polyclonal Antibody

Description
Peptide sequence: FPVFPCTLLA PPFPVLGLDS RGVGGLMNSF PPPQGHAQNP LQVGAELQSR Sequence homology: Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | MAZ |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | MAZ |
| Gene Accession No. | P56270, P56671 |
| Gene Alias | LOW QUALITY PROTEIN: myc-associated zinc finger protein; Maz; MAZI; MYC associated zinc finger protein; myc-associated zinc finger protein; MYC-associated zinc finger protein (purine-binding transcription factor); PUR1; Pur-1; purine-binding transcription factor; SAF-1; SAF-2; SAF-3; serum amyloid A activating factor 1; serum amyloid A activating factor 2; serum amyloid A-activating factor-1; Transcription factor Zif87; ZF87; Zif87; Zinc finger protein 801; zinc-finger protein, 87 kilodaltons; ZNF801 |
| Gene Symbols | MAZ |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.