Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MB67 Monoclonal Antibody (OTI1C6)
GREENER_CHOICE

Catalog No. PIMA526725
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526725 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526725 Supplier Invitrogen™ Supplier No. MA526725
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MB67 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the nuclear receptor superfamily, and is a key regulator of xenobiotic and endobiotic metabolism. The protein binds to DNA as a monomer or a heterodimer with the retinoid X receptor and regulates the transcription of target genes involved in drug metabolism and bilirubin clearance, such as cytochrome P450 family members. Unlike most nuclear receptors, this transcriptional regulator is constitutively active in the absence of ligand but is regulated by both agonists and inverse agonists.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MB67
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NR1I3
Gene Accession No. Q14994
Gene Alias AA209988; AI551208; Car; CAR1; CAR-beta; Care2; constitutive activator of retinoid response; constitutive active receptor; constitutive active response; constitutive androstane nuclear receptor variant 2; constitutive androstane nuclear receptor variant 3; constitutive androstane nuclear receptor variant 4; constitutive androstane nuclear receptor variant 5; constitutive androstane receptor; constitutive androstane receptor NR1I3; constitutive androstane receptor protein variant 1; constitutive androstane receptor protein variant 2; constitutive androstane receptor protein variant 3; constitutive androstane receptor protein variant 4; constitutive androstane receptor protein variant 5; constitutively transactivates RAREs; ESTM32; MB67; MGC150433; MGC97144; MGC97209; NR1I3; nuclear receptor; nuclear receptor constitutive active receptor; nuclear receptor subfamily 1 group I member 3; nuclear receptor subfamily 1, group I, member 3; orphan nuclear hormone receptor; orphan nuclear receptor MB67; strain Fischer nuclear receptor (CAR); Unknown (protein for MGC:137128)
Gene Symbols NR1I3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-236 of NR1I3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9970
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.