Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBOAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543193 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543193 Supplier Invitrogen™ Supplier No. PA543193
  • $589.00
    $478.65 / Each of 1

    Save $110.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Peptide sequence: AAEPQPSSLS YRTTGSTYLH PLSELLGIPL DQVNFVVCQL VALFAAFWFR Sequence homology: Horse: 100%; Human: 100%; Mouse: 91%; Pig: 93%; Rabbit: 86%; Rat: 91%.

MBOAT1 shares structural similarity with a superfamily of membrane-bound O-acetyltransferases that transfer organic compounds, usually fatty acids (e.g., cholesterol, diacylglycerol, palmitoyl), onto hydroxyl groups of membrane-embedded targets.MBOAT1 shares structural similarity with a superfamily of membrane-bound O-acetyltransferases that transfer organic compounds, usually fatty acids (e.g., cholesterol, diacylglycerol, palmitoyl), onto hydroxyl groups of membrane-embedded targets (Dauwerse et al., 2007 [PubMed 17440500]).[supplied by OMIM]. Sequence Note: The RefSeq transcript and protein were derived from genomic sequence to make the sequence consistent with the reference genome assembly. The genomic coordinates used for the transcript record were based on alignments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBOAT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Mboat1
Gene Accession No. Q6ZNC8
Gene Alias 1-acylglycerophosphoethanolamine O-acyltransferase; 1-acylglycerophosphoserine O-acyltransferase; 9130215M02Rik; BC023845; dJ434O11.1; LPE acyltransferase 1; LPEAT; Lpeat1; LPLAT; LPLAT 1; LPSAT; lyso-PE acyltransferase; lysophosphatidylethanolamine acyltransferase; lysophosphatidylethanolamine acyltransferase 1; lysophosphatidylserine acyltransferase; Lysophospholipid acyltransferase 1; lyso-PS acyltransferase; MBOAT1; membrane bound O-acyl transferase family, member 1; membrane bound O-acyltransferase domain containing 1; membrane-bound O-acyltransferase domain-containing protein 1; MGC44669; Moact1; OACT1; O-acyltransferase (membrane bound) domain containing 1; O-acyltransferase domain-containing protein 1; RGD1565521; RGD1565561
Gene Symbols Mboat1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MBOAT1 (aa 2-51).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154141
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.