Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBOAT4/Ghrelin O acyltransferase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982832
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2832 Supplier Bioss Supplier No. BS13355R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mediates the octanoylation of ghrelin at ′Ser-3′. Can use a variety of fatty acids as substrates including octanoic acid, decanoic acid and tetradecanoic acid.

Specifications

Antigen MBOAT4/Ghrelin O acyltransferase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MBOAT4
Gene Accession No. Q96T53
Gene Alias BOS_23856; FKSG89; Ghrelin O-acyltransferase; Gm171; GOAT; Mboat4; membrane bound O-acyltransferase domain containing 4; Membrane-bound O-acyltransferase domain-containing protein 4; OACT4; O-acyltransferase (membrane bound) domain containing 4; O-acyltransferase domain-containing protein 4
Gene Symbols MBOAT4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Ghrelin O acyltransferase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 619373
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.