Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143589 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143589 100 μL
PIPA5143555 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5143589 Supplier Invitrogen™ Supplier No. PA5143589
  • $538.00
    $494.65 / Each of 1

    Save $43.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

MBP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by the classic MBP gene is a major constituent of the myelin sheath of oligodendrocytes and Schwann cells in the nervous system. However, MBP-related transcripts are also present in the bone marrow and the immune system. These mRNAs arise from the long MBP gene (otherwise called Golli-MBP) that contains 3 additional exons located upstream of the classic MBP exons. Alternative splicing from the Golli and the MBP transcription start sites gives rise to 2 sets of MBP-related transcripts and gene products. The Golli mRNAs contain 3 exons unique to Golli-MBP, spliced in-frame to 1 or more MBP exons. They encode hybrid proteins that have N-terminal Golli aa sequence linked to MBP aa sequence. The second family of transcripts contain only MBP exons and produce the well characterized myelin basic proteins. This complex gene structure is conserved among species suggesting that the MBP transcription unit is an integral part of the Golli transcription unit and that this arrangement is important for the function and/or regulation of these genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MBP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene MBP
Gene Accession No. P02686, P02687, P02688, P04370, P81558
Gene Alias 20 kDa microtubule-stabilizing protein; C76307; golli mbp; Golli-mbp; Golli-MBP; myelin basic protein; Golli-Mbp; myelin basic protein; myelin basic protein S; Hmbpr; MBP; MBP S; Mbps; MGC99675; microtubule-stabilizing protein; mld; MOBP; Myelin A1 protein; myelin basic protein; myelin basic protein Golli-mbp; myelin basic protein S; myelin deficient; myelin membrane encephalitogenic protein; Myelin P1 protein; myelin-associated oligodendrocyte basic protein; R75289; shi; shiverer; Unknown (protein for IMAGE:7984318); unnamed protein product
Gene Symbols MBP
Host Species Goat
Immunogen Purified myelin basic protein isolated from bovine brain.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17196, 24547, 414286, 4155, 618684
Target Species Human, Mouse, Rat, Bovine, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.