Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva MBPTrap™ HP Prepacked Columns
GSA_VA

Catalog No. 45001542 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 1mL
1 x 5mL
5 x 1mL
5 x 5mL
Flow Rate:
<20 mL/min.
<4 mL/min.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Flow Rate
45-001-542 5 x 5mL <20 mL/min.
45-001-541 1 x 5mL <20 mL/min.
45-001-540 5 x 1mL <4 mL/min.
45-000-508 1 x 1mL <4 mL/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45-001-542 Supplier Cytiva Supplier No. 28918780
Add to Cart
Add to Cart

Yield highly pure MBP-tagged recombinant proteins eluted in concentrated form and small volumes. Cytiva Life Sciences™ MBPTrap™ HP Prepacked Columns are prepacked with Dextrin Sepharose™ High Performance.

  • Prepacked with Dextrin Sepharose tagged with maltose binding protein (MBP).
  • Binding capacity is fusion protein dependent but can be as much as 16 mg/ml media
  • Maximum pressure: 3 bar
  • Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH
  • 1 and 5mL polypropylene columns can be used either manually using a syringe or on an instrument

Media
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: >7

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffer
Product Type Dextrin Sepharose HP prepacked column
Bed Volume 1mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 5 x 5mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <20 mL/min.
Material Polypropylene
Ligand Type Dextrin
Max. Pressure 3 bar
pH Range >7
Particle Size 34μm
Support Type 6% Highly Cross-linked Agarose
Content And Storage 2° to 8°C
Column Type Ion Exchange Column
For Use With (Application) Purification of MBP tagged Proteins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.