Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBTD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735206 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5206 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5206 Supplier Proteintech Group Inc Supplier No. 260981AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MBDT1 is a putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility. MBDT1 specifically binds to monomethylated and dimethylated ′Lys-20′ on histone H4.

Specifications

Antigen MBTD1
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MBTD1
Gene Accession No. Q05BQ5, Q6P5G3
Gene Alias mbt domain containing 1, MBTD1, SA49P01
Gene Symbols MBTD1
Host Species Rabbit
Immunogen MBTD1 Fusion Protein Ag22059
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103537, 54799
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.