Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MC 1568, Tocris Bioscience™

Catalog No. 407750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-775-0 50 mg
40-771-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-775-0 Supplier R&D Systems Supplier No. 4077/50
This item is not returnable. View return policy

Selective HDAC class II (IIa) inhibitor

Selective inhibitor of class IIa histone deacetylases (HDACs). Exhibits tissue-selective inhibition between members of class II deacetylases in vivo; inhibits HDAC4 and HDAC5 in skeletal muscle and the heart without affecting HDAC3 activity. Arrests myogenesis through the stabilization of myocyte enhancer factor 2D (MEF2D)-HDAC3/4 complex. Displays no inhibition of class I HDAC activity or expression.

Chemical Identifiers

CAS 852475-26-4
Molecular Formula C17H15FN2O3
Molecular Weight (g/mol) 314.32
MDL Number MFCD16875423
InChI Key QRDAPCMJAOQZSU-UHFFFAOYSA-N
Synonym hdac-in-1, e-3-4-e-3-3-fluorophenyl-3-oxoprop-1-enyl-1-methyl-1h-pyrrol-2-yl-n-hydroxyacrylamide, 2e-3-4-1e-3-3-fluorophenyl-3-oxoprop-1-en-1-yl-1-methylpyrrol-2-yl-n-hydroxyprop-2-enamide, e-3-4-e-3-3-fluorophenyl-3-oxoprop-1-en-1-yl-1-methyl-1h-pyrrol-2-yl-n-hydroxyacrylamide, e-3-4-e-3-3-fluorophenyl-3-oxoprop-1-enyl-1-methylpyrrol-2-yl-n-hydroxyprop-2-enamide, 3-5-3-3-fluorophenyl-3-oxopropen-1-yl-1-methyl-1h-pyrrol-2-yl-n-hydroxy-2-propenamide, mc1568 inverted exclamation marky hplc, 3-4-3-3-fluorophenyl-3-oxoprop-1-enyl-1-methyl-1h-pyrrol-2-yl-n-hydroxyacrylamide, e-3-4-e-3-3-fluorophenyl-3-oxoprop-1-enyl-1-methyl-1hpyrrol-2-yl-n-hydroxyacrylamide, e-3-4-3-3-fluoro-phenyl-3-oxo-propenyl-1-methyl-1h-pyrrol-2-yl-n-hydroxy-acrylamide
PubChem CID 11381449
IUPAC Name 3-{4-[3-(3-fluorophenyl)-3-oxoprop-1-en-1-yl]-1-methyl-1H-pyrrol-2-yl}-N-hydroxyprop-2-enamide
SMILES CN1C=C(C=CC(=O)C2=CC(F)=CC=C2)C=C1C=CC(=O)NO

Specifications

Quantity 50 mg
Formula Weight 314.31
Percent Purity >96%
Chemical Name or Material MC 1568
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.