Learn More
Medchemexpress LLC HY-112786 5mg , MC-Val-Cit-PAB-MMAF CAS:863971-17-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1127865MG
Encompass_Preferred
HY-112786 5mg MC-Val-Cit-PAB-MMAF CAS: 863971-17-9 MC-Val-Cit-PAB-MMAF (Vc-MMAF) is a drug-linker conjugate for ADC with antitumor activity by using the tubulin inhibitor, MMAF, linked via cathepsin cleavable MC-Val-Cit-PAB. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9634
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.