Learn More
Medchemexpress LLC HY-128955 5mg Medchemexpress, MC-VC-PABC-Aur0101 CAS:1438849-92-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1289555MG
Encompass_Preferred
Medchemexpress, HY-128955 5mg MC-VC-PABC-Aur0101 CAS:1438849-92-3 MC-VC-PABC-Aur0101 is a drug-linker conjugate for ADC with potent antitumor activity by using Aur0101 (an auristatin microtubule inhibitor), linked via the ADC linker MC-VC-PABC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9786
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.