Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MC1R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595418
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595418 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595418 Supplier Invitrogen™ Supplier No. PA595418
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: PC12 whole cell, HEPA1-6 whole cell, HELA whole cell, A375 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

MC1R, a Melanocortin Receptor, mediates the effects of melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone (ACTH). MSH and its receptor, MC1R, are key regulators of melanogenesis. They control the relative proportions of red pheomelanin and black photoprotective eumelanin in the skin by increasing the production of eumelanin. Non-functional, truncated forms of the receptor lead to lighter coat color in animals. In contrast, constitutively active receptors lead to darker color.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MC1R
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene Mc1r
Gene Accession No. Q01726, Q01727
Gene Alias alpha melanocyte stimulating hormone receptor; CMM5; e; extension recessive yellow; MC1 receptor; MC1R; MC1-R; melanocortin 1 receptor; melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor); melanocortin receptor 1; melanocortin-1 receptor; melanocyte-stimulating hormone receptor; melanotropin receptor; MSHR; MSH-R; Mshra; SHEP2; Tob; tobacco darkening
Gene Symbols Mc1r
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human MC1 Receptor (290-317aa NAIIDPLIYAFHSQELRRTLKEVLTCSW).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102552838, 17199, 4157
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.