Learn More
CPC Scientific MCA-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Lys(DNP)-NH2 (acetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD108A
Encompass
Fluorogenic substrate for pro-memapsin-2 containing the b-secretase site of the Swedish mutation of APP. The kinetic parameters at pH 4.5 are Km = 4.5 ?M and kcat = 0.25 min-1.ONE-LETTER SEQUENCE: Mca-SEVNLDAEF-K(DNP)-NH2MOLECULAR FORMULA: C68H89N15O26MOLECULAR WEIGHT:1532.54STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1802078-32-5], SYNONYMS: Mca-(Asn,Leu)-Amyloid β/A4 Protein Precursor (667-675)-Lys(Dnp) amideRESEARCH AREA: Alzheimer's DiseaseREFERENCES: J. Ermolieff et al., Biochemistry, 39, 16263 (2000); J. Ermolieff et al., Biochemistry, 39, 12450 (2000)
Catalog No. 50-210-8732
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.