Learn More
CPC Scientific MCA-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Arg-Lys(DNP)-Arg-Arg-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific AMYD109A
Encompass
ONE-LETTER SEQUENCE: Mca-SEVNLDAEFR-K(DNP)-R-R-NH2MOLECULAR FORMULA: C85H125N27O29MOLECULAR WEIGHT:2001.10STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [438625-61-7 ], SYNONYMS: Mca-(Asn,Leu)-Amyloid β/A4 Protein Precursor (667-676)-Lys(Dnp)-Arg-Arg amideRESEARCH AREA: Alzheimer's DiseaseREFERENCES: H. Koike et al., J. Biochem., 126, 235 (1999)
Catalog No. 50-210-8734
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.