Learn More
CPC Scientific MCA-Lys-Pro-Leu-Gly-Leu-Dap(DNP)-Ala-Arg-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MMPS029A
Encompass
The N-terminal elongation of the widely used MMP FRET substrate Mca-PLGL-Dpa-AR-NH2 with a Lys yields a fluorogenic peptide with improved substrate propertiesONE-LETTER SEQUENCE: Mca-KPLGL-Dap(Dnp)-AR-NH2MOLECULAR FORMULA: C55H80N16O16MOLECULAR WEIGHT:1221.34STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [720710-69-0], RESEARCH AREA: CancerREFERENCES: U.Neumann et al., Anal. Biochem., 328, 166 (2004); M.O.Palmier and S.R.Van Doren, Anal. Biochem., 371, 43 (2007); K.Tanimoto et al., J. Dent. Res., 87, 451 (2008)
Catalog No. 50-210-9809