Learn More
Bachem Mca-Pro-Leu-OH
SDP
Supplier:  Bachem 4018898.0050
Encompass_Preferred
Mca-Pro-Leu-OH, 140430-55-3, C23H28N2O7, Mr 444.48, 50mg. MMP-2/MMP-7 Substrate Control, Fluorogenic. Standard compound for enzyme assays using Mca-containing FRET substrates. Excitation at 328 nm, emission at 393 nm.
Catalog No. NC9873830