Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mca-YVADAPK(Dnp)-OH Fluorogenic Peptide Substrate
GSA_VASDP

Catalog No. ES007 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES007 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ES007 Supplier R&D Systems™ Supplier No. ES007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Bioactivity

  • Contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2, 4-dinitrophenyl group
  • It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence
  • Purity >95% by HPLC
  • The cleavage sites by ICE and ACE-2 are the peptide bonds between Ala and Asp and between Pro and Lys, respectively

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Form Liquid
For Use With (Application) Bioactivity
Product Line R&D Systems
Quantity 1 mg
Format Supplied as a stock solution in dimethyl sulfoxide (DMSO) at a concentration of 6.19mg/mL or 4.0 mM.
Molecular Weight (g/mol) Predicted MW: 1145.2 Da
Product Type Purity: >95%, by HPLC, Source: Synthetic
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.