Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCC Rat, Mouse anti-Human, Unconjugated, Polyclonal, (secondary), Abnova™

Catalog No. 89339471 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-471 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-471 Supplier Abnova Corporation Supplier No. H00004163PW2
Add to Cart
Add to Cart

Rat, Mouse Polyclonal Antibody.

MCC Polyclonal antibody specifically detects MCC in Human samples. It is validated for IP-WB.

Specifications

Antigen MCC
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description mutated in colorectal cancers
Gene MCC
Gene Alias DKFZp762O1615, FLJ38893, FLJ46755, MCC1
Host Species Rat, Mouse
Quantity 1 Set
Primary or Secondary Primary
Gene ID (Entrez) 4163
Includes Antibody pair for IP: rabbit purified polyclonal anti-MCC (300 μg); Antibody pair for WB: mouse purified polyclonal anti-MCC (50 μg)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.