Learn More
Selleck Chemical LLC MCC950 Sodium (CP-456773 Sodium) 200mg 256373-96-3 CRID3 sodium salt
SDP
Supplier:  Selleck Chemical LLC S7809200MG
Encompass_Preferred
MCC950 Sodium (CP-456773 Sodium, CRID3 sodium salt) is a potent, selective inhibitor of NLRP3 with IC50 of 7.5 nM in BMDMs; but not the AIM2, NLRC4 or NLRP1 inflammasomes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5284
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.