Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCC950 Na, MP Biomedicals™

Catalog No. MP218358910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218358910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP218358910 Supplier MP Biomedicals, Inc Supplier No. 0218358910
Add to Cart
Add to Cart

A sodium salt that is a potent selective inhibitor of NOD-like receptor family protein3 (NLRP3) with an IC50 value of 7.5 nM in bone marrow derived from macrophages.

  • For in vivo experiments in studies regarding the nueroprotective action of individuals on a ketgenic diet using mouse models to study the effect of imflammasome activity on tumor progression with the inhibition of NLRP3
  • Study the response of palcental explants by understadning the cholesterol crystal-mediated IL-1B response
  • Soluble in water, DMSO, dimethyl formamide or ethanol
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256373-96-3
Molecular Formula C20H23N2NaO5S

Specifications

Quantity 10 mg
Formula Weight 426.5
Physical Form Crystalline Solid
Purity ≥97%
Molecular Weight (g/mol) 426.46
Chemical Name or Material MCC950 Na
Recommended Storage -20 C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.