Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525583
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525583 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525583 Supplier Invitrogen™ Supplier No. PA525583
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MCCC1 Polyclonal Antibody for Western Blot

MCCC1 is the large subunit of3-methylcrotonyl-CoA carboxylase. This enzyme functions as aheterodimer and catalyzes the carboxylation of 3-methylcrotonyl-CoAto form 3-methylglutaconyl-CoA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCCC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MCCC1
Gene Accession No. Q96RQ3, Q99MR8
Gene Alias 1810045E08Rik; 2310058B18Rik; 3-methylcrotonyl-CoA carboxylase 1; 3-methylcrotonyl-CoA carboxylase 1 {ECO:0000250; 3-methylcrotonyl-CoA carboxylase biotin-containing subunit; 3-methylcrotonyl-CoA carboxylase biotin-containing subunit {ECO:0000250; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit alpha; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit alpha {ECO:0000250; EMBL:AAH88473.1, ECO:0000312; Mcca; MCCalpha; MCCase subunit alpha; MCCase subunit alpha {ECO:0000250; MCC-B; MCCC1; mccc1 {ECO:0000312; methylcrotonoyl-CoA carboxylase 1; methylcrotonoyl-CoA carboxylase 1 (alpha); methylcrotonoyl-CoA carboxylase alpha; methylcrotonoyl-CoA carboxylase subunit alpha, mitochondrial; methylcrotonoyl-CoA carboxylase subunit alpha, mitochondrial {ECO:0000250; methylcrotonoyl-Coenzy; methylcrotonoyl-Coenzyme A carboxylase 1 (alpha); R75106; RGD:1310615}; UniProtKB:Q96RQ3}
Gene Symbols MCCC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 397-425 amino acids from the central region of human MCCC1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56922, 72039
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.