Learn More
ILEX LIFE SCIENCES LLC MCCOYS 5A P04-06500 500 ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0406500
Encompass_Preferred
PAN-Biotech McCoy’s 5A Medium (modified), w: stable Glutamine, w: 2.2 g/L NaHCO3 (500 ml). BICARBONATE: With 2.2 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With Stable Glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: McCoy's 5A Medium (modified) is a complete medium with all amino acids and vitamins. It is used for growing primary cultures. This group contains marrow cells, gingival cells, adrenal cells, spleen cells, lung cells, rat embryos and other cell types. McCoy's 5A medium was originally developed by Dr. Thomas McCoy as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A is unique in that it contains the reducing agent glutathione, bacto-peptone, and a high level of glucose.
Catalog No. 50-273-3866
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.