Learn More
StatLab MCDONALD'S GRAM STAIN KIT 1EA
SDP
Supplier:  StatLab KTMGSPT
Encompass_Preferred
This kit stains for gram positive and gram negative bacteria in less than five minutes. Stain kit includes Gentian Violet, Universal Iodine Solution, Gram's Decolorizer, Carbol Fuchsin, and Optimized Tartrazine Counter Stain.
Catalog No. NC0402042
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.