Learn More
Amsbio LLC MCF7 FFPE Cell Pellet Slide (5-pack)
Supplier:  Amsbio LLC 301002205PK
Encompass_Preferred
FFPE cell line sections are manufactured using a proprietary technology that maintains both cell morphology and preserves nucleic acids and proteins. Available from over 120 tumor cell lines, these FFPE sections make for affordable and consistent controls without compromising quality or consistency. Using cell line controls overcomes issues experienced with traditional cell or tissue-based control specimens such as intra specimen variation, lot-to-lot variations and inconsistent biomolecule quality. Also available in scroll & block formats - please enquire.
Catalog No. 50-204-1579
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.