Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam MCF7 Membrane Extract Lysate, Liquid, WB, 100 μg

Catalog No. 502678651
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-267-8651 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-267-8651 Supplier Abcam Supplier No. AB29539100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Membrane extract lysate suitable for Western blot analysis.

  • Applications: WB
  • Form: Liquid
  • Storage Buffer: 10% Glycerol (glycerin, glycerine), 2% Sodium lauryl sulfate, 1.17% 2-Mercaptoethanol, 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione, 0.1% EDTA, 0.1% HEPES, 0.1% Magnesium chloride, 0.1% Sodium chloride

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Format Tube
Form Liquid
Target Molecule MCF7 Proteins
Buffer 10% glycerol, 2% sodium lauryl sulfate, 1.17% 2-mercaptoethanol, 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione, 0.1% EDTA, 0.1% HEPES, 0.1% magnesium chloride, 0.1% sodium chloride
Formulation 10% Glycerol, 2% Sodium lauryl sulfate, 1.17% 2-Mercaptoethanol, 0.1% 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ6-benzoxathiole-1,1-dione, 0.1% EDTA, 0.1% HEPES, 0.1% Magnesium chloride, 0.1% Sodium chloride
Quantity 100 μg
For Use With (Application) Western Blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.