Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mCherry Monoclonal Antibody (16D7), Alexa Fluor™ 488
GREENER_CHOICE

Catalog No. 501138088
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-113-8088 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-113-8088 Supplier Invitrogen™ Supplier No. M11239
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

mCherry Monoclonal Antibody for Flow

MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mCherry
Applications Flow Cytometry
Classification Monoclonal
Clone 16D7
Conjugate Alexa Fluor 488
Formulation PBS with 5mM sodium azide; pH 7.5
Gene Alias mCherry
Host Species Rat
Immunogen Full-length mCherry was used as the immunogen.
Purification Method Affinity chromatography
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.