Learn More
Description
mCherry Polyclonal Antibody for Western Blot, IHC (P), ICC/IF
Specifications
Specifications
| Antigen | mCherry |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 5mM sodium azide |
| Gene Alias | mCherry |
| Host Species | Rabbit |
| Immunogen | Full length recombinant mCherry protein. |
| Purification Method | Affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
