Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mCherry Recombinant Rat Monoclonal Antibody (16D7), Alexa Fluor™ Plus 647, Invitrogen™

Catalog No. 740003TP647
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-000-3TP647 50 μL
50-274-0586 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-000-3TP647 Supplier Invitrogen™ Supplier No. 740003TP647
Add to Cart
Edge
Add to Cart

Rat Recombinant Monoclonal Antibody

mCherry Tag recombinant rat monoclonal antibody specifically recognizes mCherry Tag at both C and N-terminus of proteins. Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. This 16D7 antibody has been tested by WB and FC analysis using HEK-293E cells that were transiently transfected with mCherry-protein constructs. This may be used for WB at 1ug/mL and for FC at 0.5ug per test. Excitation: 658 nm; Emission: 675 nm; Laser: Red Laser Filtration: 0.2um post-manufacturing filtered.

MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mCherry
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone 16D7
Concentration 1.0 mg/mL
Conjugate Alexa Fluor Plus 647
Formulation Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8
Gene Alias mCherry
Host Species Rat
Immunogen Full-length protein mCherry.
Purification Method Affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.