Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCHR1 Recombinant Rabbit Monoclonal Antibody (12H4L9)

Catalog No. 702618
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-261-8 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-261-8 Supplier Invitrogen™ Supplier No. 702618
  • $462.00
    $346.65 / Each of 1

    Save $115.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Mouse, Rabbit. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

MCHR1 is a member of the G protein-coupled receptor family 1. It is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCHR1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 12H4L9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene Mchr1
Gene Accession No. P97639, Q99705
Gene Alias AW049955; G protein coupled receptor 24; G protein-coupled receptor 24; GPCR24; GPR24; Gpr24-9 protein coupled receptor; G-protein coupled receptor 24; MCH 1 receptor; MCH receptor 1; MCH1; Mch1r; MCH-1R; MCHR; MCHR1; MCH-R1; MCHR-1; melanin concentrating hormone receptor 1; melanin-concentrating hormone receptor 1; SLC1; SLC-1; Somatostatin receptor-like protein
Gene Symbols Mchr1
Host Species Rabbit
Immunogen Peptide corresponding to human MCHR1 [aa401-aa422].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2847, 83567
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.