Learn More
Epigentek Inc MCM3 Polyclonal Antibody (100 ul)
SDP
Supplier:  Epigentek Inc A68307100
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Negative elongation factor B protein (8-199AA). Tested Applications: ELISA, WB, IHC, ChIP. Reactivity: Human, Mouse. Alternative Names: Cofactor of BRCA1.
Catalog No. 50-125-0622
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.