Learn More
MilliporeSigma™ MQuant™ Carbonate Hardness Test, Freshwater/Seawater
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Format | Sample Testing Kit |
| For Use With (Application) | Determine carbonate hardness as a measure of the buffering capacity of water |
| Graduations | 1.25 °e |
| Includes | Dropper bottle, Reagent, Sample vials |
| No. of Tests | 50 |
| Quantity | 50 Tests |
| Range | 1 drop = 1.25 °e |
| Storage Requirements | Up to 3 years at 15–25°C |
| Test Method | Acidimetric, Titration with Dropper Bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.