Learn More
Specifications
Specifications
| Format | Sample Testing Kit |
| For Use With (Application) | Determine carbonate hardness as a measure of the buffering capacity of water |
| Graduations | 0.25 °e and 0.1mmol/L |
| Includes | Pipette, Reagent, Sample vials |
| No. of Tests | 300 |
| Quantity | 300 Tests |
| Range | 0.25 to 25 °e and ANC 0.1 to 7.2 mmol/L |
| Storage Requirements | Up to 3 years at 15–25°C |
| Test Method | Acidimetric, Titration with Pipette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.