Learn More
Specifications
Specifications
| Format | Sample Testing Kit |
| For Use With (Application) | Determination of formaldehyde in aqueous, disinfecting and rinsing solutions |
| Graduations | 0.10–0.25–0.4–0.6–0.8– 1.0–1.5 |
| Includes | Sliding color comparator, Reagent, Sample vials |
| No. of Tests | 100 |
| Quantity | 100 Tests |
| Range | 0.1–1.5mg/L HCHO |
| Storage Requirements | Up to 3 years at 15–25°C |
| Test Method | Colorimetric, triazole derivative indicator |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.