Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ pH Test

Catalog No. M1080270001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
400 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1080270001 400 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1080270001 Supplier MilliporeSigma™ Supplier No. 1080270001
Add to Cart
Add to Cart

Designed for highly sensitive, rapid water analysis. MilliporeSigma™ MQuant™ pH Test is a user-friendly kit for performing 400 precise pH analyses. It includes a brilliant color card to compare the reaction color of the sample with a high-quality

TRUSTED_SUSTAINABILITY

Specifications

pH Range 4.5 to 9
Format Sample Testing Kit
For Use With (Application) Water pH testing
Graduations 0.5 pH
Includes Color card comparator, Reagent, Sample vials
No. of Tests 400
Quantity 400 Tests
Storage Requirements Up to 3 years at 15–25°C
Test Method Colorimetric, mixed indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.