Learn More
MilliporeSigma™ MQuant™ Total Hardness Test Kits
Catalog No. M1080390001 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Total Hardness Test
Total Hardness Test Kit
Total Hardness Test Refill pack Reagent H-1
Range:
0.1 to 3.6 mmol/L
1 drop corresponds to 1.25°e
1 drop corresponds to 20mg/L CaCO3
1 to 100 mg/L CaCO3
Test Method:
108039 MColortest
Titrimetric w/ Titration Pipette
Titrimetric w/Dropping Bottle
No. of Tests:
50
100
200
300
600
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Range Test Method No. of Tests
M1080390001 Total Hardness Test 0.1 to 3.6 mmol/L Titrimetric w/ Titration Pipette 300
M1080470001 Total Hardness Test 1 to 100 mg/L CaCO3 Titrimetric w/ Titration Pipette 300
M1146520001 Total Hardness Test Kit 1 drop corresponds to 1.25°e Titrimetric w/Dropping Bottle 50
M1083120001 Total Hardness Test Kit 1 drop corresponds to 20mg/L CaCO3 Titrimetric w/Dropping Bottle 200
M1111220001 Total Hardness Test Refill pack Reagent H-1 - 108039 MColortest 600
M1111040001 Total Hardness Test Kit 1 drop corresponds to 1.25°e Titrimetric w/Dropping Bottle 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. M1080390001 Supplier MilliporeSigma™ Supplier No. 1080390001
Add to Cart
Add to Cart
Method: titrimetric MQuant™
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Mcolortest™ Total Hardness Test
No. of Tests 300
Range 0.1 to 3.6 mmol/L
Test Method Titrimetric w/ Titration Pipette
Type Total Hardness Test