Specifications
| For Use With (Application) | Mcolortest™ Total Hardness Test |
| No. of Tests | 300 |
| Range | 1 to 100 mg/L CaCO3 |
| Test Method | Titrimetric w/ Titration Pipette |
| Type | Total Hardness Test |